Learn More
Thermo Scientific™ TRACE™ TR-1 GC Columns
Product Code. 10374363 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
100 m
15 m
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
1.5 μm
3 μm
5 μm
Unit Size:
Each
Product Code. 10374363
Brand: Thermo Scientific™ 260A395P
Only null left
Add to basket
Add to basket
Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.
Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.32 mm
Product Line TRACE
Stationary Phase TR-1
Film Thickness 3 μm
Length (Metric) 30 m