Learn More
MP Biomedicals™ Trace Minerals for an Ultra-Clean Environment
Product Code. 11467191 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467191 100 g 100g
11477191 500 g 500g
11487191 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11467191 Supplier MP Biomedicals™ Supplier No. 02960264.1
  • ONLINE EXCLUSIVE PRICE
    118.00 EUR
    116.55 EUR / 100g
    Save 1.45 EUR
    1% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Trace Minerals is used in ultra clean environments where trace elements usually present, are absent.
  • 7-trace elements which are usually present in a open environment
  • Research animal diet supplement
  • Powdered
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
For Use With (Application) Research animal diet supplements
Quantity 100 g
Storage Requirements Room Temperature