Learn More
Thermo Scientific™ TRACE™ GC Columns for Pesticides
Product Code. 10713236 Shop All Thermo Scientific Products
Change view
Click to view available options
Stationary Phase:
Trace™ Pesticide
Trace™ Pesticide II
Trace™ Pesticide III
Trace™ Pesticide IV
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Stationary Phase Unit Size
10713236 Trace™ Pesticide III Each
10210705 Trace™ Pesticide IV Each
10773586 Trace™ Pesticide II Each
10211635 Trace™ Pesticide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10713236 Supplier Thermo Scientific™ Supplier No. 26RC142F
Estimated Shipment: 07-01-2027
Identify organophosphate and organochlorine pesticides with high performance, sensitivity and reproducibility.
Identify organophosphate and organochlorine pesticides with high performance, sensitivity, and reproducibility using the Thermo Scientific™ TRACE™ GC Columns for Pesticides. These columns are specifically designed and tested to meet the most stringent requirements for the analysis of pesticides and herbicides.
Excellent Quality and Reproducibility:
  • Low bleed decreases MS contamination
  • Innovative design facilitates use at higher temperatures
  • Column inertness results in minimal peak tailing and decreased breakdown of sensitive samples
Alternative Uses:
  • Chlorinated aromatics
  • Plant sterols
  • Pyrethroids
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Pesticides Analysis
Particle Size 0.2 μm
Film Thickness 0.25 μm
Length (Metric) 30 m
Description 30 m x 0.25 mm x 0.25 um with 5 m Guard
Diameter (Metric) 0.25 mm
USP Type G43
Stationary Phase Trace™ Pesticide III
Diameter (Metric) Inner 0.25 mm
Product Line TRACE
Max. Temperature (Metric) 280/300°C