missing translation for 'onlineSavingsMsg'
Learn More

TPT-260 Dihydrochloride, MedChemExpress

Product Code. 16475848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475848 10 mg 10mg
16471997 50 mg 50mg
16438507 5 mg 5mg
16406326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475848 Supplier Medchem Express Supplier No. HY13769A
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TPT-260 Dihydrochloride (NSC55712) is a thiophene thiourea derivative with molecule weight 260.00 in free base form; There is no formal name yet, we temporally call this molecule as TPT-260.

TPT-260 Dihydrochloride (NSC55712) is a thiophene thiourea derivative with molecule weight 260.00 in free base form

Specifications

Chemical Name or Material TPT-260 Dihydrochloride
Physical Form Solid
CAS 2076-91-7
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C8H14Cl2N4S3
Synonym NSC55712 TPU-260 Dihydrochloride
Solubility Information DMSO : 100 mg/mL (300.01 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (99.99 mM; Need ultrasonic)
SMILES NC(SCC1=CC=C(CSC(N)=N)S1)=N.Cl.Cl
Molecular Weight (g/mol) 333.32
Formula Weight 333.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.