Learn More
TPO agonist 1, MedChemExpress
Product Code. 16475717
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475717 5 mg 5mg
16459546 50 mg 50mg
16452978 10 mg 10mg
16434296 100 mg 100mg
16400016 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16475717 Supplier Medchem Express Supplier No. HY100380
MedChemExpress TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia.
TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia.
Specifications
Chemical Name or Material TPO agonist 1
CAS 1033040-23-1
Color Red
Purity Grade Notes Research
Molecular Formula C25H22N8O2
Quantity 5 mg
Solubility Information DMSO : ≥ 36 mg/mL (77.17 mM)
SMILES O=C(N(C1=CC=C(C)C(C)=C1)N=C/2C)C2=N\NC3=C(O)C(C4=CC=CC(C5=NN=NN5)=C4)=CC=C3
Molecular Weight (g/mol) 466.49
Formula Weight 466.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation