Learn More
Ansell TouchNTuff™ Disposable Powder-free Blue Nitrile Gloves
Product Code. 11853540 Shop All Ansell Products
Change view
Click to view available options
Size:
10
Large
Medium
Small
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11853540 Large Pack of 100
11843540 Medium Pack of 100
11833540 Small Pack of 100
11863540 10 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 11853540 Lieferant Ansell Lieferanten-Nr. 92665090
  • ONLINE EXCLUSIVE PRICE
    33.95 EUR
    22.50 EUR / Pack of 100
    Save 11.45 EUR
    33% Off
Alle Werbeaktionen anzeigen
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Made from an innovative nitrile compound which offers outstanding chemical and mechanical properties, comfort and dexterity.Three times the puncture resistance of latex or PVC disposable. Lighter and cooler, yet safer and more durable.
  • Three times the puncture resistance of latex or PVC disposable gloves
  • 100% nitrile; no waxes, silicone or plasticizers
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves
  • Textured finish on fingertips
  • Improved grip
  • Rolled cuff
  • Provides added protection at wrist and ensures a secure fit
TRUSTED_SUSTAINABILITY
Spezifikation
Certifications/Compliance Category III, EN374
Accelerator Free No
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 0.0055 in.
Finger Thickness (Metric) 0.14 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.0043 in.
Palm Thickness (Metric) 0.11 mm
Powdered Powder Free
Product Type Nitrile Gloves
Size Large
Sterility Non-sterile
Thickness (English) 0.14 mm
Thickness (Metric) 0.10 mm
For Use With (Application) Ideal applications chemical industry, electronics, emergency services, laboratory analysis, intricate parts handling
Quantity 100 Pk.
Series 92-665
Length (Metric) 300 mm
Mehr anzeigen Weniger anzeigen