missing translation for 'onlineSavingsMsg'
Learn More

Toremifene citrate, MedChemExpress

Product Code. 16438056
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16438056

Brand: Medchem Express HYB0005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Toremifene citrate (Z-Toremifene citrate) is a second-generation selective estrogen-receptor modulator (SERM) in development for the prevention of osteoporosis. Toremifene citrate also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.07 μM and 2.6 μM, respectively.

Toremifene citrate (Z-Toremifene citrate) is a second-generation selective estrogen-receptor modulator (SERM) in development for the prevention of osteoporosis. Toremifene citrate also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.07 μM and 2.6 μM, respectively.

Specifications

Chemical Name or Material Toremifene citrate
CAS 89778-27-8
Color White
Purity Grade Notes Research
Molecular Formula C32H36ClNO8
Quantity 100 mg
Synonym Z-Toremifene citrate NK 622 FC-1157a
Solubility Information DMSO : ≥ 100 mg/mL (167.20 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClCC/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(C)C)C=C2)\C3=CC=CC=C3.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 598.08
Formula Weight 598.08
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.