missing translation for 'onlineSavingsMsg'
Learn More

Toluene-4-sulfonic Acid Sodium Salt, Sigma-Aldrich™

Product Code. 15886680
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886680 250 g 250g
15871401 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15886680 Supplier Sigma Aldrich Supplier No. 8211250250
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-84-1

Specifications

Melting Point >300°C
Density 1.55 g/cm3 at 20°C
Molecular Formula 4-(CH3)C6H4SO3Na
MDL Number MFCD00064388
Quantity 250 g
Synonym Toluene-4-sulfonic acid sodium salt, 4-Toluenesulfonic acid sodium salt, Toluene-4-sulfonic acid sodium salt
Molecular Weight (g/mol) 194.18
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.