missing translation for 'onlineSavingsMsg'
Learn More

Toluene-4-sulfonic Acid Monohydrate, Supelco™

Product Code. 15875350
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15875350 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15875350 Supplier Supelco Supplier No. 1096130100
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Specifications

CAS 6192-52-5
Melting Point 56°C (Elimination of Water of Crystallisation)
Boiling Point 140°C/27 hPa
MDL Number MFCD00142137
Quantity 100 g
Synonym Toluene-4-sulfonic acid monohydrate, p-Toluenesulfonic acid, Tosylic acid, Tosic acid, PTSA, Toluene-4-sulfonic acid monohydrate
Molecular Weight (g/mol) 172.20 (Anhydrous Basis)
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.