missing translation for 'onlineSavingsMsg'
Learn More

Toltrazuril, TRC

Product Code. 30138074
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138074 250 mg 250mg
30135796 1 g 1g
17944155 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138074 Supplier Toronto Research Chemicals Supplier No. TRCT535850250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69004-03-1
Molecular Formula C18 H14 F3 N3 O4 S
Molecular Weight (g/mol) 425.38
Synonym 1-Methyl-3-[3-methyl-4-[4-[(trifluoromethyl)thio]phenoxy]phenyl]-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, BAY-Vi 9142, Baycox, Deltazuril, Toltrazuril, 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1-methyl-3-[3-methyl-4-[4-[(trifluoromethyl)thio]phenoxy]phenyl]-
IUPAC Name 1-methyl-3-[3-methyl-4-[4-(trifluoromethylsulfanyl)phenoxy]phenyl]-1,3,5-triazinane-2,4,6-trione
SMILES CN1C(=O)NC(=O)N(C1=O)c2ccc(Oc3ccc(SC(F)(F)F)cc3)c(C)c2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H14F3N3O4S/c1-10-9-11(24-16(26)22-15(25)23(2)17(24)27)3-8-14(10)28-12-4-6-13(7-5-12)29-18(19,20)21/h3-9H,1-2H3,(H,22,25,26)
Formula Weight 425.0657
Percent Purity >95%
Chemical Name or Material Toltrazuril
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.