Learn More
Toltrazuril, MedChemExpress
Product Code. 16480798
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480798 5 g 5g
16402079 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16480798 Supplier Medchem Express Supplier No. HYB0175
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Toltrazuril (BAY-i 9142) is an antiprotozoal agent that acts upon Coccidia parasites.
Toltrazuril (BAY-i 9142) is an antiprotozoal agent that acts upon Coccidia parasites.
Specifications
Chemical Name or Material Toltrazuril
Physical Form Solid
CAS 69004-03-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C18H14F3N3O4S
For Use With (Application) COVID-19-immunoregulation
Synonym BAY-i 9142
Solubility Information DMSO : 130 mg/mL (305.61 mM; Need ultrasonic)
SMILES O=C(N(C1=CC=C(OC2=CC=C(SC(F)(F)F)C=C2)C(C)=C1)C(N3)=O)N(C)C3=O
Molecular Weight (g/mol) 425.38
Formula Weight 425.38
Percent Purity 98.65%
Grade Research