missing translation for 'onlineSavingsMsg'
Learn More

Tolperisone Hydrochloride, Tolperisone Hydrochloride, TRC

Product Code. 17913475
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913475 1 g 1g
17901365 50 mg 50mg
17973165 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913475 Supplier Toronto Research Chemicals Supplier No. TRCT5354751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C16 H23 N O . Cl H
Molecular Weight (g/mol) 281.82
Synonym 2-Methyl-1-(4-methylphenyl)-3-(1-piperidinyl)-1-propanone Hydrochloride (1:1), 1-Propanone, 2-methyl-1-(4-methylphenyl)-3-(1-piperidinyl)-, hydrochloride (9CI), Propiophenone, 2,4'-dimethyl-3-piperidino-, hydrochloride (7CI,8CI), 1-Piperidino-2-methyl-3-(4-methylphenyl)propan-3-one hydrochloride, 2,4'-Dimethyl-3-piperidinopropiophenone hydrochloride, Abbsa, Arantoick, Atmosgen, Besnoline, Biocalm, Isocalm, Kineorl, Menopatol, Metosomin, Midocalm, Minacalm, Muscalm, Mydocalm, N 553, Naismeritin, Tolfree, Tolisartine, Tolperisone hydrochloride, Tolpidol
IUPAC Name 2-methyl-1-(4-methylphenyl)-3-piperidin-1-ylpropan-1-one;hydrochloride
SMILES Cl.CC(CN1CCCCC1)C(=O)c2ccc(C)cc2

Specifications

CAS 3644-61-9
Quantity 1 g
InChI Formula InChI=1S/C16H23NO.ClH/c1-13-6-8-15(9-7-13)16(18)14(2)12-17-10-4-3-5-11-17;/h6-9,14H,3-5,10-12H2,1-2H3;1H
Formula Weight 281.1546
Percent Purity >95%
Chemical Name or Material Tolperisone Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.