Learn More
Tolmetine, TRC
Product Code. 30133914
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133914 100 mg 100mg
30012881 10 mg 10mg
30008270 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133914 Supplier Toronto Research Chemicals Supplier No. TRCM332943100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26171-23-3
Molecular Formula C15 H15 N O3
Molecular Weight (g/mol) 257.28
Synonym 1H-Pyrrole-2-acetic acid, 1-methyl-5-(4-methylbenzoyl)-, Pyrrole-2-acetic acid, 1-methyl-5-p-toluoyl- (8CI), 1-Methyl-5-(4-methylbenzoyl)-1H-pyrrole-2-acetic acid, 1-Methyl-5-p-toluoylpyrrole-2-acetic acid, 5-[(p-Tolyl)carbonyl]-1-methylpyrrole-2-acetic acid, McN 2559, Tolmetin, Tolmetine
IUPAC Name 2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetic acid
SMILES Cc1ccc(cc1)C(=O)c2ccc(CC(=O)O)n2C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H15NO3/c1-10-3-5-11(6-4-10)15(19)13-8-7-12(16(13)2)9-14(17)18/h3-8H,9H2,1-2H3,(H,17,18)
Formula Weight 257.1052
Chemical Name or Material Tolmetin