Learn More
Tolfenamic acid, 99+%, Thermo Scientific™
NSAID that activates calcium-activated potassium channels
90.65 EUR - 150.00 EUR
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15463869
SDS View Documents
Thermo Scientific Alfa Aesar
J61256.06
ONLINE EXCLUSIVE PRICE
101.00 EUR
90.65 EUR / 5g
Save 10.35 EUR
10% Off
View alternative products
Save Up to 
»
15473869
SDS View Documents
Thermo Scientific Alfa Aesar
J61256.09
10g
150.00 EUR
10g
View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water (slightly), acetone (∼10 mg/ml), DMSO (52 mg/ml at 25°C), methanol (∼10 mg/ml) and ethanol (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.