Learn More
Tolfenamic acid, 99+%, Thermo Scientific™
NSAID that activates calcium-activated potassium channels
Brand:  Thermo Scientific Alfa Aesar J61256.06
 View more versions of this product
Product Code. 15463869
  • ONLINE EXCLUSIVE PRICE
    101.00 EUR
    90.65 EUR / 5g
    Save 10.35 EUR
    10% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water (slightly), acetone (∼10 mg/ml), DMSO (52 mg/ml at 25°C), methanol (∼10 mg/ml) and ethanol (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Promotions
Promotions