Learn More
Tolbutamide, 98%, Thermo Scientific™
46.30 EUR - 615.00 EUR
Chemical Identifiers
| CAS | 64-77-7 |
|---|---|
| Molecular Formula | C12H18N2O3S |
| Molecular Weight (g/mol) | 270.347 |
| MDL Number | MFCD00027169 |
| InChI Key | JLRGJRBPOGGCBT-UHFFFAOYSA-N |
| Synonym | tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon |
| PubChem CID | 5505 |
| ChEBI | CHEBI:27999 |
| IUPAC Name | 1-butyl-3-(4-methylphenyl)sulfonylurea |
| SMILES | CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11460287
|
Thermo Scientific Alfa Aesar
B21698.09 |
10g |
46.30 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11470287
|
Thermo Scientific Alfa Aesar
B21698.18 |
50g |
168.00 EUR
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11480287
|
Thermo Scientific Alfa Aesar
B21698.30 |
250g |
615.00 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsTolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.
Solubility
Soluble in chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| C12H18N2O3S | |
| MFCD00027169 | |
| tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon | |
| CHEBI:27999 | |
| CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C |
Specifications
| 64-77-7 | |
| MFCD00027169 | |
| 14,9507 | |
| Soluble in chloroform. | |
| CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C | |
| 270.347 | |
| CHEBI:27999 | |
| 98% |
| C12H18N2O3S | |
| 1984428 | |
| tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon | |
| JLRGJRBPOGGCBT-UHFFFAOYSA-N | |
| 1-butyl-3-(4-methylphenyl)sulfonylurea | |
| 5505 | |
| 270.35 | |
| Tolbutamide |
Safety and Handling
GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
missing translation for 'einecsNumber' : 200-594-3
missing translation for 'rtecsNumber' : YS4550000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Tolbutamide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Tolbutamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.