missing translation for 'onlineSavingsMsg'
Learn More

Tolbutamide, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar B21698.09


Quantity 10g

 View more versions of this product

Product Code. 11460287

  • 46.30 EUR / 10g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

64-77-7
270.347
JLRGJRBPOGGCBT-UHFFFAOYSA-N
5505
1-butyl-3-(4-methylphenyl)sulfonylurea
C12H18N2O3S
MFCD00027169
tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon
CHEBI:27999
CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C

Specifications

64-77-7
MFCD00027169
1984428
tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon
JLRGJRBPOGGCBT-UHFFFAOYSA-N
1-butyl-3-(4-methylphenyl)sulfonylurea
5505
270.35
Tolbutamide
C12H18N2O3S
10g
14,9507
Soluble in chloroform.
CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C
270.347
CHEBI:27999
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tolbutamide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Tolbutamide

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.