missing translation for 'onlineSavingsMsg'
Learn More

TMS, MedChemExpress

Product Code. 16487596
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487596

Brand: Medchem Express HY19340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TMS ((E)-2,3',4,5'-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. TMS shows a lesser extent inhibitory effect on CYP1A1 (IC5050 =3.1 μM). TMS is a methylated derivative of resveratrol and has anti-cancer activity.

TMS ((E)-2,3',4,5'-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. TMS shows a lesser extent inhibitory effect on CYP1A1 (IC5050 =3.1 μM). TMS is a methylated derivative of resveratrol and has anti-cancer activity.

Specifications

Chemical Name or Material TMS
CAS 24144-92-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H20O4
Quantity 100 mg
Synonym (E)-2,3',4,5'-tetramethoxystilbene
Solubility Information DMSO : ≥ 50 mg/mL (166.47 mM)
SMILES COC1=CC=C(/C=C/C2=CC(OC)=CC(OC)=C2)C(OC)=C1
Molecular Weight (g/mol) 300.35
Formula Weight 300.35
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.