Learn More
Tizanidine-d4 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Tizanidine-d4 Hydrochloride |
| CAS | 1188263-51-5 |
| Molecular Formula | C9H5D4Cl2N5S |
| Quantity | 10 mg |
| Synonym | 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl-d4)-2,1,3-benzothiadiazol-4-amine Hydrochloride |
| InChI Formula | InChI=1S/C9H8ClN5S.ClH/c10-5-1-2-6-8(15-16-14-6)7(5)13-9-11-3-4-12-9;/h1-2H,3-4H2,(H2,11,12,13);1H/i3D2,4D2; |
| SMILES | ClC1=C(NC2=NC([2H])([2H])C([2H])([2H])N2)C3=NSN=C3C=C1.Cl |
| IUPAC Name | 5-chloro-N-(4,4,5,5-tetradeuterio-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride |
| Molecular Weight (g/mol) | 294.2 |
| Formula Weight | 293.0207 |