Learn More
BRAND™ Titrette™ Digital Bottle-Top Burette
Product Code. 10175324 Shop All Brand Products
Change view
Click to view available options
Accuracy:
18 μL (0.07 %)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Unit Size
10175324 18 μL (0.07 %) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10175324 Supplier BRAND™ Supplier No. 4760451
  • ONLINE EXCLUSIVE PRICE
    1330.00 EUR
    1026.19 EUR / Pack of 1
    Save 303.81 EUR
    22% Off
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Bulk Quote Request
The first bottletop burette to meet class A error limits for glass burettes
  • Stands out for easy titration, a compact design, excellent precision, an exchangeable dosing unit, and an optional PC interface
  • You can use the Titrette™ to titrate quickly and reliably, even with restricted space and independent of available power connections - in the laboratory, in production, or in the field
  • Incorporates a simple and effective design for bottle attachment
  • Separate buttons for on/off, pause, and CLEAR to delete the display and select function
  • Non-slip hand wheels and a smooth-running precision gear set for fast or drip titration make handling reliable and especially simple
  • TRUSTED_SUSTAINABILITY
    Specifications
    Accuracy 18 μL (0.07 %)
    Coefficient of Variation 6 μL (0.025%)
    Volume (Metric) Not applicable as device works according to Soxhlet-Henkel
    Interface Without
    Model According to Soxhlet-Henkel