Learn More
SI Analytics™ Titration Tip
Product Code. 10663222 Shop All SI Analytics Products
Change view
Click to view available options
Diameter (Metric):
0.7 mm
0.8 mm
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10663222 0.8 mm Each
10287781 0.7 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10663222 Supplier SI Analytics™ Supplier No. 285214265
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
SI Analytics™ Titration Tip is designed to be attached to a titrator to increase precision in titration application.
  • Tip for titrator
  • For use with TITRONIC™ Series and TitroLine™ Series
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.8 mm
Type Titration Tip
Length (Metric) 180 mm