missing translation for 'onlineSavingsMsg'
Learn More

Titrant for Sulfur Dioxide Mini Titrator, Hanna Instruments™

Product Code. 11612410
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11612410 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
*The HI84100 Sulfur Dioxide Mini Titrator has been discontinued. The new version is the HI84500 Sulfur Dioxide Mini Titrator for Wine Analysis which has its own set of reagents.
Product Code. 11612410 Supplier Hanna Instruments Supplier No. HI8410050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
*The HI84100 Sulfur Dioxide Mini Titrator has been discontinued. The new version is the HI84500 Sulfur Dioxide Mini Titrator for Wine Analysis which has its own set of reagents.

Used for low range determination of sulfur dioxide in wine using the HI84100 Sulfur Dioxide Mini Titrator for Wine Analysis.* Hanna Instruments™ Titrant for Sulfur Dioxide Mini Titrator is pre-standardized, making titrations fast, accurate and easy to perform.

The HI84100 mini titrator is designed to determine free and total sulfur dioxide levels in wine by potentiometric titration. The HI3148B ORP electrode is used to detect the endpoint. The HI84100 can be used with all wines, including red, which are difficult to test using traditional methods due to the difficulty to observe a distinctive color change when the endpoint is reached.

  • Pre-standardized titrant
  • Clearly marked expiration date and lot number
  • Air tight bottle with tamper-proof seal

Specifications

Color Colorless
CAS 5-6-7758
CAS Min % 1%
CAS Max % 3%
Density 1.01
Solubility Information Soluble in water
Odor Odorless
Packaging Air tight bottle with tamper-proof seal
pH 12
Quantity 100 mL
Flash Point >60°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.