Learn More
Titanium Standard Metal Solution 1000ppm, for ICP, Solution HCl 2M, Fisher Chemical
Product Code. 10625882 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10625882 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10625882 Supplier Fisher Chemical Supplier No. J/8331/08
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging HDPE plastic bottle
Physical Form Liquid
MDL Number 11264
Color Purple
Quantity 500 mL
compliance-icons
Product Identifier
  • Titanium solution 1000 ppm
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid