Learn More
Honeywell™ Titan Work Positioning Lanyard
Product Code. 12624983
Change view
Click to view available options
Length (Metric):
1.8 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12624983 1.8 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12624983 Supplier Honeywell™ Supplier No. 1008287
  • ONLINE EXCLUSIVE PRICE
    73.50 EUR
    59.14 EUR / Pack of 1
    Save 14.36 EUR
    19% Off
Estimated Shipment: 08-02-2027
Ideal positioning lanyard for a full-range of applications including construction and general maintenance.
  • User-friendly fall protection
  • Meets industry and construction compliance requirements
  • Ideal for those who work at height in a safe and supported hands-free position
  • Recommended for use in construction
  • Critical tool for worker protection that combines mobility with unsurpassed comfort
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 3, AQ: ISO 9001/2000, EC Certificate: 333, EC Attestation: 0082/490/160/10/10/0284 , Conforms to EN 358
Includes Loop and 2 twist-lock carabiners (21 mm)
Length (Metric) 1.8 m
Material Polyamide