Learn More
Bel Art™ Tissue Culture Flask Racks
Product Code. 11722016
Change view
Click to view available options
Positions:
5
12
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Unit Size
11722016 12 Each
11732016 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11722016 Supplier Bel Art™ Supplier No. F189700001
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 05-11-2026
Racks prevent the domino effect of flasks tipping one over the other.
  • These polypropylene racks hold 25cc, 75cc, 50 to 75mL, or 250 to 260 mL or similar sized tissue culture flasks in a vertical position
  • Rectangular holes in the top and center plates stabilize the flasks preventing tipping
  • Easily organize flasks to improve processing and to avoid errors in collecting used media, media change, washing of cell monolayer, trypsinization and seeding of cells
  • Grips on each end make rack handling easy and allow for safe transport of flasks to the incubator
  • Steam autoclavable at 121°C (250°F)
Specifications
Autoclavable Autoclavable
Color White
Holds Flasks
Material Polypropylene
Positions 12
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) 50 to 75 mL Flat-sided Tissue Culture Flask
Temperature (Metric) Range 121°C
Dimensions (L x W x H) 23 x 13.7 x 10.7 cm (9.1 x 5.4 x 4.2 in.)
Length (Metric) 230 mm
Height (English) 4.2 in.
Product Type Flask Rack
Shelves 3 Tiers
Height (Metric) 10.7 cm
Width (Metric) 137 mm