Learn More
Tipelukast, TRC
Specifications
Specifications
| Chemical Name or Material | Tipelukast |
| CAS | 125961-82-2 |
| Molecular Formula | C29H38O7S |
| Quantity | 2.5 mg |
| Synonym | 4-[6-Acetyl-3-[3-[(4-acetyl-3-hydroxy-2-propylphenyl)thio]propoxy]-2-propylphenoxy]-butanoic Acid, 4-[6-Acetyl-3-[3-(4-acetyl-3-hydroxy-2-propylphenylthio)propoxy]-2-propylphenoxy]butyric Acid, KCA 757, MN 001 |
| InChI Formula | InChI=1S/C29H38O7S/c1-5-9-23-25(14-12-22(20(4)31)29(23)36-16-7-11-27(32)33)35-17-8-18-37-26-15-13-21(19(3)30)28(34)24(26)10-6-2/h12-15,34H,5-11,16-18H2,1-4H3,(H,32,33) |
| SMILES | OC(=O)CC(=O)OCC1[C@H](O)C(O)C(O)[C@@H](O1)Oc2cc3c(c(O)c2)C(=O)C(=CO3)c4ccc(O)cc4 |
| IUPAC Name | 4-[6-acetyl-3-[3-(4-acetyl-3-hydroxy-2-propylphenyl)sulfanylpropoxy]-2-propylphenoxy]butanoic acid |
| Molecular Weight (g/mol) | 530.67 |
| Formula Weight | 530.2338 |