Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 10μL
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Pack of 19200
Description
- Manufactured to stringent specifications under strict process controls
- Assembled via automation to ensure consistent performance and quality
- Certified RNase-/DNase-free and nonpyrogenic
- 384-well racks of non-filtered, non-sterile pipette 10μL tips
- Compatible with Agilent™/Velocity VPrep™ and Bravo 384ST, 96ST and 16ST head
Order Info
Minimum Order Quantity is 10 cases.
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Clean Claims | DNase, RNase Free |
| For Use With (Application) | Non-pyrogenic |
| Format | Rack |
| Tip Style | Automation, Round |
| For Use With (Equipment) | Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST |
| Material | Polypropylene |