Learn More
Lovibond™ Tintometer™ SensoDirect 150 All-In-One Handheld Meter, pH/Redox
Description
- Includes:
- Instrument
- Batteries
- pH electrode
- Temperature probe
- Redox electrode
- pH buffer set with pH 4.0 and pH 7
- Instruction manual
- Guarantee sheet in case
- Features
- pH/Redox, Temperature °C/°F
- All in one
- Real time data logger
- Protective casing
- RS 232 / USB
Specifications
Specifications
| Accuracy (pH) | 0 to 14 pH, ±0.02 pH + 2 digits |
| Accuracy (mV) | 0 to 1999mV, 1mV |
| Accuracy (Temperature) | ±0.8°C (1.5°F) |
| Type | SensoDirect 150 Set pH/Redox |
| Dimensions (L x W x H) | 220 x 120 x 40 mm |
| For Use With (Application) | Measures pH/Redox, dissolved oxygen and conductivity/TDS |
| pH Range | 0 to 14 |
| Range (Conductivity) | 0 to 200.0 µS |
| Range (mV) | mV to 1999mV to 1999mV |
| Range (TDS) | 0 to 132,000ppm |
| Show More |