missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Eriochrome Hardness Test Powder, 100 tests

Product Code. 15331715
Change view
Click to view available options
No. of Tests:
100
Unit Size:
20g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
15331715 100 20g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15331715 Supplier Lovibond™ Supplier No. 462950
  • ONLINE EXCLUSIVE PRICE
    109.00 EUR
    107.40 EUR / 20g
    Save 1.60 EUR
    1% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers

  • Eriochrome Hardness Test Powder
  • 100 tests

Specifications

For Use With (Equipment) Test kits, colorimeters and spectrophotometers
No. of Tests 100
Type Eriochrome Hardness Test Powder
compliance-icons compliance-icons
Product Identifier
  • 20GR HARDNESS ERIOCHROME POWDER .
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: boric acid, disodiumtetraborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.