missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Chloride Dihydrate, Supelco™

Product Code. 15835050
Change view
Click to view available options
Quantity:
250 g
2.5 kg
Unit Size:
2.5kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15835050 2.5 kg 2.5kg
15825050 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15835050 Supplier Supelco Supplier No. 1078142500
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

For analysis (max. 0.000001% Hg) EMSURE™

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 10025-69-1

Tekniske data

Boiling Point 623°C/1013 hPa (anhydrous substance)
Melting Point 38°C (anhydrous substance)
Quantity 2.5 kg
MDL Number MFCD00149863
Synonym Tin(II) chloride dihydrate, Tin dichloride, dihydrate, Stannous chloride, Tin(II) chloride dihydrate
Molecular Weight (g/mol) 225.65
Grade Research
Density 2.71 g/cm3 at 20°C
Chemical Name or Material Tin(II) Chloride Dihydrate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.