missing translation for 'onlineSavingsMsg'
Learn More

Timolol Maleate, Timolol Maleate, TRC

Product Code. 30133827
Change view
Click to view available options
Quantity:
100 mg
2.5 g
10 g
Unit Size:
100mg
10g
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133827 2.5 g 2.50g
30132605 10 g 10g
18073020 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133827 Supplier Toronto Research Chemicals Supplier No. TRCT4437102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13 H24 N4 O3 S . C4 H4 O4
Molecular Weight (g/mol) 432.49
Synonym Timolol Maleate, 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (2S)-, (2Z)-2-butenedioate (1:1), 2-Propanol, 1-(tert-butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]-, (-)-, maleate (1:1) (salt) (8CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (2S)-, (2Z)-2-butenedioate (1:1) (salt) (9CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (S)-, (Z)-2-butenedioate (1:1) (salt), 1,2,5-Thiadiazole, 2-propanol deriv., (-)-Timolol maleate, (S)-(-)-Timolol maleate, (S)-Timolol hydrogen maleate, (S)-Timolol maleate, Aquanil, Betim, Betime, Blocadren, Blocanol, Istalol, L-Timolol maleate, MK 950, Optimol, Proflax, Rysmon TG, Temserin, Tenopt, Timabak, Timacar, Timacor, Timolet, Timolol LA, Timolol hydrogen maleate, Timomann, Timoptic, Timoptol, Timoptol XE, Timorom, WP 934, l-Timolol maleate
IUPAC Name (Z)-but-2-enedioic acid;(2S)-1-(tert-butylamino)-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-2-ol
SMILES CC(C)(C)NC[C@H](O)COc1nsnc1N2CCOCC2.OC(=O)\C=C/C(=O)O

Specifications

CAS 26921-17-5
Quantity 2.5 g
InChI Formula InChI=1S/C13H24N4O3S.C4H4O4/c1-13(2,3)14-8-10(18)9-20-12-11(15-21-16-12)17-4-6-19-7-5-17;5-3(6)1-2-4(7)8/h10,14,18H,4-9H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t10-;/m0./s1
Formula Weight 432.1679
Percent Purity >95%
Chemical Name or Material Timolol Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.