Learn More
Timapiprant sodium, MedChemExpress
Product Code. 16452567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Packungsgröße:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
16452567 50 mg 50mg
16429057 5 mg 5mg
16416468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16452567 Lieferant Medchem Express Lieferanten-Nr. HY15342A
 Restricted Item
Estimated Shipment: 30-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
MedChemExpress Timapiprant sodium (OC000459 sodium) is a potent, selective, and orally active D prostanoid receptor 2 (DP2, also known as CRTH2) antagonist. Timapiprant sodium (OC000459 sodium) potently displaces [3H] PGD2 from human recombinant DP2 (Ki=13 nM), rat recombinant DP2 (Ki=3 nM), and human native DP2 (Ki=4 nM). Timapiprant sodium (OC000459 sodium) inhibits mast cell activation of Th2 lymphocytes and eosinophils.
Timapiprant sodium (OC000459 sodium) is a potent, selective, and orally active D prostanoid receptor 2 (DP2, also known as CRTH2) antagonist. Timapiprant sodium (OC000459 sodium) potently displaces [3H] PGD2 from human recombinant DP2 (Ki=13 nM), rat recombinant DP2 (Ki=3 nM), and human native DP2 (Ki=4 nM). Timapiprant sodium (OC000459 sodium) inhibits mast cell activation of Th2 lymphocytes and eosinophils.
Spezifikation
Chemical Name or Material Timapiprant sodium
CAS 950688-14-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H16FN2NaO2
Quantity 50 mg
Synonym OC000459 sodium
Solubility Information DMSO : 100 mg/mL (270.01 mM; Need ultrasonic)
SMILES O=C(O[Na])CN1C2=CC=C(F)C=C2C(CC3=NC4=C(C=CC=C4)C=C3)=C1C
Molecular Weight (g/mol) 370.35
Formula Weight 370.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Mehr anzeigen Weniger anzeigen