missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiafenacil Metabolite M-56, TRC
Specifications
Specifications
| Chemical Name or Material | Tiafenacil Metabolite M-56 |
| Analyte or Component Names | Tiafenacil Metabolite M-56 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H9 Cl F4 N2 O5 S |
| Synonym | 2-((2-Chloro-5-(2,6-dioxo-4-(trifluoromethyl)-3,6-dihydropyrimidin-1(2H)-yl)-4-fluorophenyl)sulfinyl)propanoic acid |
| InChI Formula | InChI=1S/C14H9ClF4N2O5S/c1-5(12(23)24)27(26)9-3-8(7(16)2-6(9)15)21-11(22)4-10(14(17,18)19)20-13(21)25/h2-5H,1H3,(H,20,25)(H,23,24) |
| Solution Type | Neat |
| SMILES | CC(C(=O)O)S(=O)c1cc(N2C(=O)NC(=CC2=O)C(F)(F)F)c(F)cc1Cl |
| IUPAC Name | 2-[2-chloro-5-[2,4-dioxo-6-(trifluoromethyl)-1H-pyrimidin-3-yl]-4-fluorophenyl]sulfinylpropanoic acid |
| Industry Type | Water Analysis |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?