Learn More
Thymolphthalein, Supelco™
Product Code. 15877650
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877650 100 g 100g
15815110 5 g 5g
15867650 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15877650 Supplier Supelco Supplier No. 1081750100
 Restricted Item
Estimated Shipment: 18-11-2026
Indicator ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 125-20-2
Specifications
Melting Point 253°C
MDL Number MFCD00005909
Synonym Thymolphthalein, Thymolphthalein
Quantity 100 g
Molecular Weight (g/mol) 430.54
Grade Research
Chemical Name or Material Thymolphthalein