Learn More
Threaded Adapter (2 in.) (Stainless Steel), Supelco™
Product Code. 15882391
Change view
Click to view available options
Quantity:
1 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15882391 1 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15882391 Supplier Supelco Supplier No. 9671009003
 Restricted Item
Estimated Shipment: 09-11-2026