Learn More
SCAT™ Thread Adapter, Type 22
Product Code. 10314370 Shop All SCAT Europe Products
Change view
Click to view available options
Closure Size:
R 2 in. BSP/G 2 in., S60/61
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
10314370 R 2 in. BSP/G 2 in., S60/61 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10314370 Supplier SCAT™ Supplier No. 107023
  • ONLINE EXCLUSIVE PRICE
    32.75 EUR
    30.35 EUR / Pack of 1
    Save 2.40 EUR
    7% Off
Estimated Shipment: 19-11-2026
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Closure Size R 2 in. BSP/G 2 in., S60/61
Closure Color Yellow
Liner Material Polypropylene
Product Type Thread Adapter