Learn More
Thorium Standard: Th @ 1000 μg/mL in 5% HNO3, VHG™
Product Code. 17997979
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17997979 50 mL 50mL
17968769 500 mL 500mL
17909389 250 mL 250mL
17937629 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17997979 missing translation for 'mfr' VHG Labs missing translation for 'supplierNo' VHGPTHN50
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulierung: Dieses Produkt enthält einen Ausgangsstoff für Explosivstoffe gemäß der Verordnung (EG) Nr. 1148/2019. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Thorium Standard: Th @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material Thorium
CAS 7440-29-1
Methods ICP/ICP-MS
Analyte or Component Names Thorium
Shipping Condition Room Temperature
Molecular Formula Th
Certifications/Compliance ISO 17034
Synonym Th
InChI Formula InChI=1S/Th
SMILES [Th]
IUPAC Name thorium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 232.038
Quantity 50 mL
Formula Weight 232.038 g/mol
Mehr anzeigen Weniger anzeigen