missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carboxaldoxime, 98%, Thermo Scientific™

Product Code. p-7046095
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11317906

Brand: Thermo Scientific Alfa Aesar L13114.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42466-50-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00151853
InChI Key CPCZUXSHORSJIZ-ZZXKWVIFSA-N
Synonym thiophene-3-carbaldehyde oxime, thiophene-3-carboxaldoxime, hydroxyimino-3-thienylmethane, thiophene-3-carboxaldehyde oxime, e-n-thiophen-3-ylmethylidene hydroxylamine, 3-thiophenecarboxaldehyde oxime, 3-thiophenecarboxaldehyde, oxime, e-n-hydroxy-1-thiophen-3-yl methanimine, 3-thiophenecarboxaldehyde, oxime, e-9ci
PubChem CID 9582358
IUPAC Name (NE)-N-(thiophen-3-ylmethylidene)hydroxylamine
SMILES C1=CSC=C1C=NO

Specifications

Melting Point 121°C to 122°C
Quantity 1 g
Beilstein 108818
Formula Weight 127.17
Percent Purity 98%
Chemical Name or Material Thiophene-3-carboxaldoxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.