CAS RN 42466-50-2
CAS RN 42466-50-2
IUPAC Name:
N-[(thiophen-3-yl)methylidene]hydroxylamine
Synonyms:
N-(thiophen-3-ylmethylidene)hydroxylamine
Molecular Weight (g/mol):
127.16
Molecular Formula:
C5H5NOS
InChi Key:
CPCZUXSHORSJIZ-UHFFFAOYSA-N
SMILES:
ON=CC1=CSC=C1
1
–
1
of
1
results
1
Thiophene-3-carboxaldoxime, 98%, Thermo Scientific™
CAS: 42466-50-2 Molecular Formula: C5H5NOS Molecular Weight (g/mol): 127.161 MDL Number: MFCD00151853 InChI Key: CPCZUXSHORSJIZ-ZZXKWVIFSA-N Synonym: thiophene-3-carbaldehyde oxime,thiophene-3-carboxaldoxime,hydroxyimino-3-thienylmethane,thiophene-3-carboxaldehyde oxime,e-n-thiophen-3-ylmethylidene hydroxylamine,3-thiophenecarboxaldehyde oxime,3-thiophenecarboxaldehyde, oxime,e-n-hydroxy-1-thiophen-3-yl methanimine,3-thiophenecarboxaldehyde, oxime, e-9ci PubChem CID: 9582358 IUPAC Name: (NE)-N-(thiophen-3-ylmethylidene)hydroxylamine SMILES: C1=CSC=C1C=NO