missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonitrile, 98%

Product Code. p-7081786
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453914 25 g 25g
11463914 100 g 100g
11473914 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453914 Supplier Thermo Scientific Alfa Aesar Supplier No. A11930.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1003-31-2 | C5H3NS | 109.146 g/mol

Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-31-2
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00005416
InChI Key CUPOOAWTRIURFT-UHFFFAOYSA-N
Synonym 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
PubChem CID 66087
IUPAC Name thiophene-2-carbonitrile
SMILES C1=CSC(=C1)C#N

Specifications

Density 1.171
Boiling Point 191°C to 193°C
Flash Point 81°C (178°F)
Odor Unpleasant
Refractive Index 1.5645
Quantity 25 g
UN Number UN3276
Beilstein 107035
Solubility Information Slightly miscible with water.
Formula Weight 109.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thiophene-2-carbonitrile
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.