missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioacetazone, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Thioacetazone
Specifications
Specifications
| Chemical Name or Material | Thioacetazone |
| CAS | 104-06-3 |
| Analyte or Component Names | Thioacetazone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 N4 O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Acetamide, N-[4-[[2-(aminothioxomethyl)hydrazinylidene]methyl]phenyl]-, Acetamide, N-[4-[[(aminothioxomethyl)hydrazono]methyl]phenyl]- (9CI), Acetanilide, 4'-formyl-, 4'-(thiosemicarbazone) (8CI), Acetanilide, 4'-formyl-, thiosemicarbazone (6CI), 4-Acetylaminobenzaldehyde thiosemicarbazone, 4207RP, 4'-Formylacetanilide thiosemicarbazone, Aktivan, Ambathizon, Amithiozone, Amitiozon, Antib, Benthiozone, Benzothiozane, Benzothiozon, Berculon A, Berkazon, Conteben, Diasan, Diazan, Diazane, Domakol, Ilbion, Livazone, Mirizone Neustab, Mivizon, Myvizone, NSC 3550, Neotibil, Neustab, Novakol, Nuclon argentinian, Panrone, Parazone, SQ 2321, Sdt 1041, Seroden, Siocarbazone, Tb I, Tb I/698, Tebalon, Tebecure, Tebemar, Tebesone I, Tebethion, Tebethione, Tebezon, Thiacetazone, Thiacetone, Thibon, Thibone, Thioacetazon, Thioacetazone, Thiocarbazil, Thiocarbazyl, Thiomicid, Thionicid, Thioparamizon, Thioparamizone, Thiosemicarbazone, Thiosemicarbazone (pharmaceutical), Thiotebesin, Thiotebezin, Thiotebicina, Thizone, Tiacetazon, Tibicur, Tibion, Tibione, Tibizan, Tibon, Tibone, Tioacetazon, Tioatsetazon, Tiobicina, Tiocarone, Tiosecolo, Tubercazon, Tubigal, Tubin, p-Acetamidobenzaldehyde thiosemicarbazone, p-Acetylaminobenzaldehyde thiosemicarbazone |
| InChI Formula | InChI=1S/C10H12N4OS/c1-7(15)13-9-4-2-8(3-5-9)6-12-14-10(11)16/h2-6H,1H3,(H,13,15)(H3,11,14,16)/b12-6+ |
| SMILES | CC(=O)Nc1ccc(\C=N\NC(=S)N)cc1 |
| IUPAC Name | N-[4-[(E)-(carbamothioylhydrazinylidene)methyl]phenyl]acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction