Learn More
Epredia™ Thinline Slotted Cassettes in Tube Packs
Product Code. 16384106 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16384106 White Pack of 750
16384096 Beige Pack of 750
16394096 Blue Pack of 750
16304106 Green Pack of 750
16314106 Gray Pack of 750
16324106 Lavender Pack of 750
16334106 Light Green Pack of 750
16344106 Lilac Pack of 750
16354106 Orange Pack of 750
16364106 Pink Pack of 750
16374106 Tan Pack of 750
16394106 Yellow Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 16384106 Leverantör Epredia™ Leverantörsnummer B851736WH
  • ONLINE EXCLUSIVE PRICE
    158.00 EUR
    122.75 EUR / Pack of 750
    Save 35.25 EUR
    22% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifikationer
Pore Style Slots
Closure Included Yes
Quantity 750/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color White
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Slotted Cassette
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.