missing translation for 'onlineSavingsMsg'
Learn More

Thifluzamide, TRC

Product Code. 30007586
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007586 1 g 1g
30012859 500 mg 500mg
17924875 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007586 Supplier Toronto Research Chemicals Supplier No. TRCT3447051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130000-40-7
Molecular Formula C13 H6 Br2 F6 N2 O2 S
Molecular Weight (g/mol) 528.06
Synonym 5-Thiazolecarboxamide, N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)- (9CI, ACI), N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-5-thiazolecarboxamide (ACI), 2',6'-Dibromo-2-methyl-4'-trifluoromethoxy-4-trifluoromethyl-1,3-thiazole-5-carboxyanilide, Greatam, MON 24000, Pulsor, Thifluzamide
IUPAC Name N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxamide
SMILES Cc1nc(c(s1)C(=O)Nc2c(Br)cc(OC(F)(F)F)cc2Br)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H6Br2F6N2O2S/c1-4-22-10(12(16,17)18)9(26-4)11(24)23-8-6(14)2-5(3-7(8)15)25-13(19,20)21/h2-3H,1H3,(H,23,24)
Formula Weight 525.8421
Percent Purity >95%
Chemical Name or Material Thifluzamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.