missing translation for 'onlineSavingsMsg'
Learn More

Thifluzamide, MedChemExpress

Product Code. 16452296
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452296 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452296 Supplier Medchem Express Supplier No. HYB2004
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Thifluzamide, a broad-spectrum succinate dehydrogenase inhibitor (SDHI) fungicide, has been widely used in the controlling of a variety of fungal diseases in rice fields.

Thifluzamide, a broad-spectrum succinate dehydrogenase inhibitor (SDHI) fungicide, has been widely used in the controlling of a variety of fungal diseases in rice fields.

Specifications

Chemical Name or Material Thifluzamide
CAS 130000-40-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C13H6Br2F6N2O2S
Quantity 500 mg
Solubility Information DMSO : ≥ 29 mg/mL (54.92 mM)
SMILES O=C(C1=C(C(F)(F)F)N=C(C)S1)NC2=C(Br)C=C(OC(F)(F)F)C=C2Br
Molecular Weight (g/mol) 528.06
Formula Weight 528.06
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.