Learn More
Thiane, TRC
Product Code. 30455253
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
5g
Product Code. Quantity unitSize
30455253 5 g 5g
30453144 10 g 10g
30450170 25 g -
3 options
Product Code. 30455253
Brand: Toronto Research Chemicals TRCT3441455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1613-51-0
Formula Weight 102.0503
InChI Formula InChI=1S/C5H10S/c1-2-4-6-5-3-1/h1-5H2
IUPAC Name thiane
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.2
SMILES C1CCSCC1
Synonym NSC 9459, Pentamethylene Sulfide, Penthiophane, Tetrahydro-2H-thiopyran, Tetrahydrothiapyran, Tetrahydrothiopyran, Thiacyclohexane
Chemical Name or Material Thiane
Specifications
Quantity 5 g