accessibility menu, dialog, popup

UserName

CAS RN 1613-51-0

IUPAC Name: thiane
Synonyms: thiane
Molecular Weight (g/mol): 102.2
Molecular Formula: C5H10S
InChi Key: YPWFISCTZQNZAU-UHFFFAOYSA-N
SMILES: C1CCSCC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)