Learn More
Thiamine pyrophosphate, MedChemExpress
Product Code. 16455566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16455566 Supplier Medchem Express Supplier No. HY113076
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
MedChemExpress Thiamine pyrophosphate is the coenzyme form of Vitamin B1 and is a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex.
Thiamine pyrophosphate is the coenzyme form of Vitamin B1 and is a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex.
Specifica
Chemical Name or Material Thiamine pyrophosphate
CAS 154-87-0
Color White
Purity Grade Notes Research
Molecular Formula C12H19ClN4O7P2S
Quantity 100 mg
Solubility Information H2O : 41.67 mg/mL (90.44 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OP(OP(OCCC1=C(C)[N+](CC2=CN=C(C)N=C2N)=CS1)(O)=O)(O)=O.[Cl-]
Molecular Weight (g/mol) 460.77
Formula Weight 460.77
Percent Purity 98.75%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Vedi altri risultati Mostra meno risultati
Research purposes only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.