Learn More
Thiamine monochloride, MedChemExpress
Artikelnummer. 16412019
Encompass_Preferred
Ansicht ändern
Click to view available options
Quantity:
1 g
Packungsgröße:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16412019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16412019 Lieferant Medchem Express Lieferanten-Nr. HYA0100
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
MedChemExpress Thiamine monochloride (Vitamin B1) is an essential vitamin that plays an important role in cellular production of energy from ingested food and enhances normal neuronal actives.
Thiamine monochloride (Vitamin B1) is an essential vitamin that plays an important role in cellular production of energy from ingested food and enhances normal neuronal actives.
Spezifikation
Chemical Name or Material Thiamine monochloride
CAS 59-43-8
Color White
Purity Grade Notes Research
Molecular Formula C12H17ClN4OS
Quantity 1 g
Synonym Vitamin B1
Solubility Information H2O : ≥ 100 mg/mL (332.44 mM) ∣DMSO : 1 mg/mL (3.32 mM; Need ultrasonic)
SMILES CC1=C(CCO)SC=[N+]1CC2=CN=C(C)N=C2N.[Cl-]
Molecular Weight (g/mol) 300.81
Formula Weight 300.81
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Mehr anzeigen Weniger anzeigen