Learn More
Thiamethoxam, TRC
Product Code. 30138330
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138330 1 g 1g
17989209 500 mg 500mg
17917689 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138330 Supplier Toronto Research Chemicals Supplier No. TRCT3441801G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 153719-23-4
Molecular Formula C8 H10 Cl N5 O3 S
Molecular Weight (g/mol) 291.71
Synonym 4H-1,3,5-Oxadiazin-4-imine, 3-[(2-chloro-5-thiazolyl)methyl]tetrahydro-5-methyl-N-nitro-, 3-[(2-Chloro-5-thiazolyl)methyl]tetrahydro-5-methyl-N-nitro-4H-1,3,5-oxadiazin-4-imine, A 97565N, A 9765N, Actara, Actara 25WG, Actara 2GR, Adage, Adage 5FS, CGA 293343, Centric, Cruiser, Cruiser 350FS, Cruiser 5FS, Cruiser 600FS, Cruiser A 9765, Diacloden, FS 350, FS 600, FS 600 (insecticide), Flagship, Meridian, Optigard, Platinum (insecticide), Platinum 240SC, Thiamethoxam, WS 70, WS 70 (insecticide), Xamox
IUPAC Name N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-5-methyl-1,3,5-oxadiazinan-4-ylidene]nitramide
SMILES CN1COCN(Cc2cnc(Cl)s2)C1=N[N+](=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3
Formula Weight 291.0193
Percent Purity >95%
Chemical Name or Material Thiamethoxam
Grade HPLC