Learn More
Quickfit™ Thermoplastic Polyester Tubing Clamps
Product Code. 12316357
Change view
Click to view available options
For Use With:
Up to 10mm O.D. tubing
Up to 14mm O.D. tubing
Up to 4.5mm O.D. tubing
Up to 6mm O.D. tubing
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
12316357 Up to 10mm O.D. tubing Pack of 5
11961528 Up to 14mm O.D. tubing Pack of 5
12704406 Up to 4.5mm O.D. tubing Pack of 5
11332025 Up to 6mm O.D. tubing Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12316357 Supplier Quickfit Supplier No. KT10
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
For use with flexible rubber and plastic tubing
  • Provides fine flow control of liquids and gases
  • Easy one handed adjustment: just roll serated wheel along sloped floor
  • Manufactured from thermoplastic polyester
TRUSTED_SUSTAINABILITY
Specifications
Material Thermoplastic Polyester
Diameter (English) Outer 0.39 in.
Diameter (Metric) Outer 10 mm
For Use With Up to 10mm O.D. tubing
For Use With (Application) Used with flexible Rubber and Plastic tubing
Flexibility Flexible
Product Type Tubing Clamp